
Tramadol Related Compound A

Synonym(s):
trans-Tramadol hydrochloride, (±)-trans-2-[(Dimethylamino)methyl]-1-(3-methoxyphenyl)cyclohexanol hydrochloride
Empirical Formula (Hill Notation):
C16H25NO2 · HCl
CAS Number:
Molecular Weight:
299.84
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2