Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TissueFab
®
bioink
Products
Brand
Sigma-Aldrich (18)
Color
pH Value
Physical Form
Quality Segment
Application
Particle Size (µm)
Polymer Composition
Polymer Type
Sterilization Method
Filter Products
Showing 1-18 of 18
Compare
Product Number
Description
Pricing
905429
TissueFab
®
bioink ,
(Gel)ma -UV/365 nm
Expand
View Pricing
918741
TissueFab
®
bioink ,
(Gel)ma -VIS/405nm, low endotoxin
Expand
View Pricing
906913
TissueFab
®
bioink ,
Alg(Gel)ma -Vis/525 nm
Expand
View Pricing
905410
TissueFab
®
bioink ,
Alg(Gel)ma -UV/365 nm
Expand
View Pricing
906905
TissueFab
®
bioink ,
Sacrificial
Expand
View Pricing
919926
TissueFab
®
bioink ,
Crosslinking solution, low endotoxin
Expand
View Pricing
920983
TissueFab
®
bioink ,
(GelAlg)ma -UV/365 nm
Expand
View Pricing
921610
TissueFab
®
bioink ,
(GelAlg)ma -Vis/405 nm
Expand
View Pricing
922862
TissueFab
®
bioink ,
(GelAlgHA)ma -Vis/405 nm
Expand
View Pricing
927201
TissueFab
®
bioink ,
(GelHA)ma -Vis/405nm, low endotoxin
Expand
View Pricing
919624
TissueFab
®
bioink ,
(GelHA)ma -Vis/405 nm
Expand
View Pricing
934178
TissueFab
®
bioink ,
(GelHAHep)MA Vis/405 nm, low endotoxin
Expand
View Pricing
934437
TissueFab
®
bioink ,
(GelHep)MA Vis/ 405nm, low endotoxin
Expand
View Pricing
919632
TissueFab
®
bioink ,
(GelHA)ma -UV/365 nm
Expand
View Pricing
920975
TissueFab
®
bioink ,
(GelAlgHA)ma -UV/365 nm
Expand
View Pricing
927252
TissueFab
®
bioink ,
(GelAlgHA)MA Vis/405 nm, low endotoxin
Expand
View Pricing
930016
TissueFab
®
bioink ,
Facile Curable GelHA, Ionic
Expand
View Pricing
927228
TissueFab
®
bioink ,
(GelAlg)ma-Vis/405nm, low endotoxin
Expand
View Pricing