
Tioconazole for system suitability

Synonym(s):
Tioconazole, 1-[2-[(2-Chloro-3-thienyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole, Thioconazole
Empirical Formula (Hill Notation):
C16H13Cl3N2OS
CAS Number:
Molecular Weight:
387.71
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing