Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tin(II) chloride
Synonym(s):
Stannous chloride, Tin(II) chloride
Linear Formula:
SnCl
2
CAS Number:
7772-99-8
Molecular Weight:
189.62
EC Number:
231-868-0
Products
Product Category
salts (3)
Brand
Sigma-Aldrich (5)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Anhydrous (2)
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
208256
Tin(II) chloride,
reagent grade, 98%
Expand
View Pricing
8.18150
Tin(II) chloride,
anhydrous for synthesis
Expand
View Pricing
908924
Tin(II) chloride,
free-flowing, Redi-Dri
™
, reagent grade, 98%
Expand
View Pricing
452335
Tin(II) chloride,
anhydrous, powder, ≥99.99% trace metals basis
Expand
View Pricing
204722
Tin(II) chloride,
≥99.99% trace metals basis
Expand
View Pricing