
Ticagrelor impurity E

Synonym(s):
(1R,2S)-2-(3,4-Difluorophenyl)cyclopropan-1-amine (R)-mandelate
Empirical Formula (Hill Notation):
C9H9F2N · C8H8O3
CAS Number:
Molecular Weight:
321.32
Products
Formula Weight
Manufacturer
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing