Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tetraethylammonium chloride
Synonym(s):
TEA chloride
Linear Formula:
(C
2
H
5
)
4
N(Cl)
CAS Number:
56-34-8
Molecular Weight:
165.70
EC Number:
200-267-5
Products
Product Category
detergents (1)
proteins (1)
Brand
Sigma-Aldrich (2)
Biological Source
Color
Formula Weight
Gene Alias
Physical Form
Product Line
Purity
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
T2265
Tetraethylammonium chloride,
≥98% (titration)
Expand
View Pricing
86614
Tetraethylammonium chloride,
BioUltra, ≥99.0% (AT)
Expand
View Pricing
86616
Tetraethylammonium chloride,
for electrochemical analysis, ≥99.0%
Expand
View Pricing