Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
tert
-Butylhydroquinone
Synonym(s):
2-(1,1-Dimethylethyl)-1,4-benzenediol, NSC 4972, tert-Butylhydroquinone
Linear Formula:
(CH
3
)
3
CC
6
H
3
-1,4-(OH)
2
CAS Number:
1948-33-0
Molecular Weight:
166.22
EC Number:
217-752-2
Products
Product Category
preservatives (1)
Brand
Sigma-Aldrich (2)
Supelco (2)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
Monomer Type
API Family
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
112941
tert
-Butylhydroquinone,
97%
Expand
View Pricing
8.41424
tert
-Butylhydroquinone,
for synthesis
Expand
View Pricing
PHR3512
tert
-Butylhydroquinone,
certified reference material, pharmaceutical secondary standard
Expand
View Pricing
07293
tert
-Butylhydroquinone,
analytical standard
Expand
View Pricing