Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
tert
-Butyl hydroperoxide solution
Synonym(s):
1,1-Dimethylethyl hydroperoxide, 2-Hydroperoxy-2-methylpropane, TBHP
Linear Formula:
(CH
3
)
3
COOH
CAS Number:
75-91-2
Molecular Weight:
90.12
EC Number:
200-915-7
Products
Brand
Sigma-Aldrich (4)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
416665
tert
-Butyl hydroperoxide solution,
5.0-6.0 M in decane
Expand
View Pricing
458139
tert
-Butyl hydroperoxide solution,
70 wt. % in H
2
O
Expand
View Pricing
418064
tert
-Butyl hydroperoxide solution,
5.0-6.0 M in nonane
Expand
View Pricing
03-4991
tert
-Butyl hydroperoxide solution,
CP
Expand
View Pricing