Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Terpinen 4-ol
Synonym(s):
4-Carvomenthenol, 4-Terpinenol
Empirical Formula (Hill Notation):
C
10
H
18
O
CAS Number:
562-74-3
Molecular Weight:
154.25
EC Number:
209-235-5
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Application
Reference Material Type
Special Grade
primary reference standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
03900590
Terpinen 4-ol,
primary reference standard
Expand
View Pricing