Skip to Content
Merck
Terconazole European Pharmacopoeia (EP) Reference Standard

Terconazole

Terconazole European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(+-)-1-{4-[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-ylmethoxy]phenyl}-4-isopropylpiperazine, Triaconazole, (±)​-​Terconazole, (±)-1-{4-[cis-2-(2,4-Dichlorophenyl)-2-(1H-1,2,4-triazol-1-ylmethyl)-1,3-dioxolan-4-ylmethoxy]phenyl}-4-isopropylpiperazine, cis-​1-​[4-​[[2-​(2,​4-​Dichlorophenyl)​-​2-​(1H-​1,​2,​4-​triazol-​1-​ylm ethyl)​-​1,​3-​dioxolan-​4-​yl]​methoxy]​phenyl]​-​4-​(1-​methylethyl-​piperazine, Terconazole
Empirical Formula (Hill Notation):
C26H31Cl2N5O3
CAS Number:
Molecular Weight:
532.46

Products

Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique