Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tantalum/Tungsten
Products
Manufacturer
Physical Form
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
GF46578729
Tantalum/Tungsten,
foil, Ta 95%/W 5%, thickness 1.0 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF92990357
Tantalum/Tungsten,
foil, Ta97.5%/W 2.5%, thickness 1.0 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF41744145
Tantalum/Tungsten,
foil, Ta 90%/W 10%, thickness 1.0 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF33518156
Tantalum/Tungsten,
foil, Ta 95%/W 5%, thickness 1.0 mm, size 50 × 50 mm, temper as rolled
Expand
View Pricing
GF57170189
Tantalum/Tungsten,
foil, Ta 90%/W 10%, thickness 0.40 mm, size 50 × 50 mm, temper annealed
Expand
View Pricing
GF65637557
Tantalum/Tungsten,
foil, thickness 0.40 mm, Ta90%/W 10%, size 100 × 100 mm, temper annealed
Expand
View Pricing
GF94715736
Tantalum/Tungsten,
foil, thickness 1.0 mm, Ta97.5%/W 2.5%, size 100 × 100 mm, temper as rolled
Expand
View Pricing
GF04347514
Tantalum/Tungsten,
foil, Ta 90%/W 10%, thickness 1.0 mm, size 100 × 100 mm, temper as rolled
Expand
View Pricing