Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Strontium chloride hexahydrate
Synonym(s):
Strontium chloride hexahydrate
Linear Formula:
SrCl
2
· 6H
2
O
CAS Number:
10025-70-4
Molecular Weight:
266.62
EC Number:
233-971-6
Products
Product Category
salts (4)
Brand
Sigma-Aldrich (3)
Supelco (1)
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
255521
Strontium chloride hexahydrate,
ACS reagent, 99%
Expand
View Pricing
1.07865
Strontium chloride hexahydrate,
for analysis EMSURE
®
ACS
Expand
View Pricing
204463
Strontium chloride hexahydrate,
99.995% trace metals basis
Expand
View Pricing
28-5250
Strontium chloride hexahydrate,
SAJ first grade, ≥98.0%
Expand
View Pricing