Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless steel - AISI 304l
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-20 of 34
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF65383106
Stainless steel - AISI 304l,
rod, Fe/Cr18%/Ni10%, 2.5 mm diameter, length 1000 mm
Expand
View Pricing
GF75485199
Stainless steel - AISI 304l,
wire, 0.6 mm diameter, Fe/Cr18%/Ni10%, length 10 m, temper annealed
Expand
View Pricing
GF99987440
Stainless steel - AISI 304l,
wire, Fe/Cr18%/Ni10%, 0.5 mm diameter, length 20 m, temper hard
Expand
View Pricing
GF02225102
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 3.0 mm, size 25 × 25 mm
Expand
View Pricing
GF16969270
Stainless steel - AISI 304l,
rod, Fe/Cr18%/Ni 10%, 25 mm diameter, length 200 mm, temper annealed
Expand
View Pricing
GF13436423
Stainless steel - AISI 304l,
rod, Fe/Cr18%/Ni 10%, 2.5 mm diameter, length 500 mm
Expand
View Pricing
GF97629610
Stainless steel - AISI 304l,
rod, 2.5 mm diameter, Fe/Cr18%/Ni10%, length 100 mm
Expand
View Pricing
GF12226632
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 3.0 mm, size 50 × 50 mm
Expand
View Pricing
GF46134413
Stainless steel - AISI 304l,
wire, Fe/Cr18%/Ni 10%, 0.5 mm diameter, length 50 m, temper hard
Expand
View Pricing
GF71222093
Stainless steel - AISI 304l,
wire, 0.6 mm diameter, Fe/Cr18%/Ni10%, length 50 m, temper annealed
Expand
View Pricing
GF86018362
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni10%, thickness 0.5 mm, size 300 × 300 mm, temper annealed
Expand
View Pricing
GF13109532
Stainless steel - AISI 304l,
rod, Fe/Cr18%/Ni 10%, 25 mm diameter, length 100 mm, temper annealed
Expand
View Pricing
GF33317131
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 3.0 mm, size 100 × 100 mm
Expand
View Pricing
GF41817854
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 3.0 mm, size 75 × 75 mm
Expand
View Pricing
GF13674595
Stainless steel - AISI 304l,
rod, Fe/Cr18%/Ni 10%, 2.5 mm diameter, length 200 mm
Expand
View Pricing
GF21000519
Stainless steel - AISI 304l,
wire, Fe/Cr18%/Ni 10%, 0.6 mm diameter, length 20 m, temper annealed
Expand
View Pricing
GF52713356
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 0.5 mm, size 50 × 50 mm, temper annealed
Expand
View Pricing
GF05401123
Stainless steel - AISI 304l,
wire, Fe/Cr18%/Ni 10%, 0.5 mm diameter, length 10 m, temper hard
Expand
View Pricing
GF29069556
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 0.15 mm, length 0.5 m, coil width 12 mm
Expand
View Pricing
GF35363404
Stainless steel - AISI 304l,
foil, Fe/Cr18%/Ni 10%, thickness 0.5 mm, size 150 × 150 mm, temper annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2