Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Specac
®
Omni Cell windows
Products
Manufacturer
Package Size
Technique
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z804088
Specac
®
Omni Cell windows,
CaF2, 41mm x 23mm x 4mm, for Omni Cell
Expand
View Pricing
Z803855
Specac
®
Omni Cell windows,
NaCl, 41mm x 23mm x 4mm, for Omni Cell
Expand
View Pricing
Z803979
Specac
®
Omni Cell windows,
KBr, 41mm x 23mm x 4mm, for Omni Cell
Expand
View Pricing
Z804193
Specac
®
Omni Cell windows,
BaF
2
, 41mm x 23mm x 4mm, for Omni Cell
Expand
View Pricing
Z804304
Specac
®
Omni Cell windows,
ZnSe, 41mm x 23mm x 2mm, for Omni Cell
Expand
View Pricing
Z804428
Specac
®
Omni Cell windows,
KRS-5, 41mm x 23mm x 4mm, for Omni Cell
Expand
View Pricing
Z805076
Specac
®
Omni Cell windows,
Polyethylene, 41mm x 23mm x 3mm, for Omni Cell
Expand
View Pricing
Z804754
Specac
®
Omni Cell windows,
Silica (IR), 41mm x 23mm x 2mm, for Omni Cell
Expand
View Pricing