Skip to Content
Merck
Soybean flour Type I

Soybean flour

Soybean flour Type I

Products

Product Category
Brand
Biological Source
Physical Form
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing