Skip to Content
Merck
Sodium taurocholate hydrate ≥97.0% (TLC)

Sodium taurocholate hydrate

Sodium taurocholate hydrate ≥97.0% (TLC)
Synonym(s):
Taurocholic acid sodium salt
Empirical Formula (Hill Notation):
C26H44NNaO7S · xH2O
CAS Number:
Molecular Weight:
537.68 (anhydrous basis)
EC Number:

Products

Product Category
Biological Source
Formula Weight
Physical Form
Purity
Tissue
Functional Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing