Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium phosphate dibasic heptahydrate
Synonym(s):
Disodium hydrogen phosphate heptahydrate, Disodium phosphate
Linear Formula:
Na
2
HPO
4
· 7H
2
O
CAS Number:
7782-85-6
Molecular Weight:
268.07
EC Number:
237-707-0
Products
Product Category
salts (2)
buffers (1)
Feature
Formula Weight
pH Value
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Special Grade
ACS reagent (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S9390
Sodium phosphate dibasic heptahydrate,
ACS reagent, 98.0-102.0%
Expand
View Pricing
431478
Sodium phosphate dibasic heptahydrate,
≥99.99% trace metals basis
Expand
View Pricing
S2429
Sodium phosphate dibasic heptahydrate,
meets USP testing specifications
Expand
View Pricing