Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium carbonate solution
Linear Formula:
Na
2
CO
3
CAS Number:
497-19-8
Molecular Weight:
105.99
Products
Brand
Supelco (2)
Formula Weight
Physical Form
Product Line
Quality Segment
Reaction Type
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1.60310
Sodium carbonate solution,
c(Na₂CO₃), 0.05 M (0.1 N), ready-to-use volumetric solution for titration, Titripur
®
Expand
View Pricing
1.09940
Sodium carbonate solution,
c(Na₂CO₃) = 0.05 mol/l (0.1 N), concentrate for preparing 1000 ml volumetric solution for titration, Titrisol
®
Expand
View Pricing