Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Silicon carbide - reaction bonded
Products
Manufacturer
Physical Form
Length (mm)
OD (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF77520832
Silicon carbide - reaction bonded,
rod, 10.0 mm diameter, length 100 mm
Expand
View Pricing
GF84138318
Silicon carbide - reaction bonded,
sheet, thickness 5.0 mm, size 50 × 50 mm, condition ground
Expand
View Pricing
GF98436561
Silicon carbide - reaction bonded,
tube, outside diameter 10.0 mm, length 45 mm, wall thickness 2.8 mm
Expand
View Pricing
GF87641387
Silicon carbide - reaction bonded,
tube, outside diameter 15.0 mm, wall thickness 4.0 mm, length 150 mm
Expand
View Pricing
GF55172965
Silicon carbide - reaction bonded,
rod, 5.0 mm diameter, length 100 mm
Expand
View Pricing
GF40639035
Silicon carbide - reaction bonded,
tube, outside diameter 10.0 mm, length 100 mm, wall thickness 2.8 mm
Expand
View Pricing
GF61122608
Silicon carbide - reaction bonded,
sheet, thickness 6.0 mm, condition ground, size 50 × 50 mm
Expand
View Pricing