Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Shikimic acid
Synonym(s):
(3R,4S,5R)-(−)-3,4,5-Trihydroxy-1-cyclohexenecarboxylic acid
Empirical Formula (Hill Notation):
C
7
H
10
O
5
CAS Number:
138-59-0
Molecular Weight:
174.15
EC Number:
205-334-2
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Supelco (2)
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
pharmaceutical secondary standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S5375
Shikimic acid,
≥99%, suitable for ligand binding assays
Expand
View Pricing
PHR3878
Shikimic acid,
certified reference material, pharmaceutical secondary standard
Expand
View Pricing
69686
Shikimic acid,
analytical standard
Expand
View Pricing