Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Septa, Thermoseal
®
, pkg 100
Products
Product Category
septa (2)
Brand
Supelco (2)
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
27193
Septa, Thermoseal
®
, pkg 100,
High temperature PTFE/silicone, diam. × thickness 13 mm × 0.060 in., for use with 7mL vial
Expand
View Pricing
27195
Septa, Thermoseal
®
, pkg 100,
High temperature PTFE/silicone, diam. × thickness 18 mm × 0.060 in., for use with 22mL vial
Expand
View Pricing