Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
SealPlate
®
ColorTab
™
films
Products
Product Category
sealing films and foils (6)
Color
Manufacturer
Length (cm)
Length (mm)
Package Size
Sterility
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
Z721417
SealPlate
®
ColorTab
™
films,
green
Expand
View Pricing
Z721468
SealPlate
®
ColorTab
™
films,
blue
Expand
View Pricing
Z721425
SealPlate
®
ColorTab
™
films,
lavender
Expand
View Pricing
Z721476
SealPlate
®
ColorTab
™
films,
assorted colors
Expand
View Pricing
Z721433
SealPlate
®
ColorTab
™
films,
orange
Expand
View Pricing
Z721441
SealPlate
®
ColorTab
™
films,
red
Expand
View Pricing