Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Screw cap, solid top with aluminum liner, pkg 100
Products
Product Category
caps (4)
Brand
Supelco (3)
Closure Type
Color
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
27142
Screw cap, solid top with aluminum liner, pkg 100,
black phenolic solid cap, aluminum liner, for use with 4 mL vial with 13-425 thread
Expand
View Pricing
27092-U
Screw cap, solid top with aluminum liner, pkg 100,
black phenolic solid cap, aluminum liner, for use with 2 mL vial (standard opening), thread for 8-425
Expand
View Pricing
27153
Screw cap, solid top with aluminum liner, pkg 100,
black phenolic solid cap, aluminum liner, for use with 7 mL vial with 15-425 thread, 15-425 thread
Expand
View Pricing
27164
Screw cap, solid top with aluminum liner, pkg 100,
black phenolic solid cap, aluminum liner, thread for 18-400, for use with 15 mL vial
Expand
View Pricing