Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Safranin O
Synonym(s):
Basic Red 2, Cotton Red, Gossypimine, Safranin T, Safranin Y or A
Empirical Formula (Hill Notation):
C
20
H
19
ClN
4
CAS Number:
477-73-6
Molecular Weight:
350.84
EC Number:
207-518-8
Products
Product Category
stains (3)
Brand
Sigma-Aldrich (3)
Color
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
S8884
Safranin O,
Dye content, ≥80%, certified by the Biological Stain Commission, powder
Expand
View Pricing
84120
Safranin O,
for microscopy (Bact., Bot., Hist.), indicator (pH 0.3-1.0)
Expand
View Pricing
S2255
Safranin O,
Dye content ≥85 %, Powder
Expand
View Pricing