Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ruthenium(III) chloride hydrate
Synonym(s):
Ruthenium trichloride
Linear Formula:
RuCl
3
· xH
2
O
CAS Number:
14898-67-0
Molecular Weight:
207.43 (anhydrous basis)
EC Number:
233-167-5
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (4)
Formula Weight
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Reagent Type
Core
Special Grade
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
206229
Ruthenium(III) chloride hydrate,
ReagentPlus
®
Expand
View Pricing
463779
Ruthenium(III) chloride hydrate,
99.98% trace metals basis
Expand
View Pricing
931578
Ruthenium(III) chloride hydrate,
≥99.9% trace metals basis
Expand
View Pricing
84050
Ruthenium(III) chloride hydrate,
38.0-42.0% Ru basis
Expand
View Pricing