Skip to Content
Merck
RuPhos 95%

RuPhos

RuPhos 95%
Synonym(s):
2-Dicyclohexylphosphino-2′,6′-diisopropoxybiphenyl
Empirical Formula (Hill Notation):
C30H43O2P
CAS Number:
Molecular Weight:
466.64

Products

Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing