
Ropinirole Related Compound A

Synonym(s):
4-(2-Hydroxyethyl)indolin-2-one, 1,3-Dihydro-4-(2-hydroxyethyl)-2H-indol-2-one
Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2