Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ribonuclease A from bovine pancreas
Synonym(s):
Pancreatic Ribonuclease, RNAsea, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase
CAS Number:
9001-99-4
EC Number:
232-646-6
Products
Product Category
enzymes (2)
Brand
Sigma-Aldrich (9)
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Application
Technique
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
R4875
Ribonuclease A from bovine pancreas,
Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Expand
View Pricing
R6513
Ribonuclease A from bovine pancreas,
for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Expand
View Pricing
R5125
Ribonuclease A from bovine pancreas,
Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Expand
View Pricing
R4642
Ribonuclease A from bovine pancreas,
(Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Expand
View Pricing
R5503
Ribonuclease A from bovine pancreas,
Type I-AS, 50-100 Kunitz units/mg protein
Expand
View Pricing
R5500
Ribonuclease A from bovine pancreas,
Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Expand
View Pricing
R5000
Ribonuclease A from bovine pancreas,
Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Expand
View Pricing
R5250
Ribonuclease A from bovine pancreas,
Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein
Expand
View Pricing
83834
Ribonuclease A from bovine pancreas,
4×cryst., ~70 U/mg (acc. to Kunitz)
Expand
View Pricing