Skip to Content
Merck
(R)-(+)-4-Methoxy-α-methylbenzylamine ChiPros®, produced by BASF, 99%

(R)-(+)-4-Methoxy-α-methylbenzylamine

(R)-(+)-4-Methoxy-α-methylbenzylamine ChiPros®, produced by BASF, 99%
Synonym(s):
(R)-(+)-1-(4-Methoxyphenyl)ethylamine
Empirical Formula (Hill Notation):
C9H13NO
CAS Number:
Molecular Weight:
151.21

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing