Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
RapidMicroBio Perforated Lid
Products
Product Category
accessories (5)
Brand
Millipore (5)
Agency
Diameter (mm)
Compatibility
Application
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.17316
RapidMicroBio Perforated Lid,
hole D 0.6 mm, aluminum lid, for use with MAS-100 NT
®
Expand
View Pricing
1.19166
RapidMicroBio Perforated Lid,
400 hole x 0.7 mm, anodized aluminum, for MAS-100 NT
®
series, MAS-100 VF
®
and MAS-100 Eco
®
suitable for Growth Direct cassettes
Expand
View Pricing
1.17315
RapidMicroBio Perforated Lid,
hole D 0.6 mm, aluminum lid, for use with MAS-100 NT
®
, for use with MAS-100 VF
®
Expand
View Pricing
1.17314
RapidMicroBio Perforated Lid,
hole D 0.6 mm, stainless steel lid, for use with MAS-100
®
Iso
Expand
View Pricing
1.17318
RapidMicroBio Perforated Lid,
hole D 0.7 mm, no. of holes, 400 autoclavable, aluminum lid, for use with MAS-100 NT
®
, for use with MAS-100 VF
®
Expand
View Pricing