Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Quercitrin
Synonym(s):
3,3′,4′,5,7-Pentahydroxyflavone 3-rhamnoside, 3-
O
-Rhamnosylquercetin, Quercetin 3-rhamnoside, Quercimelin, Quercitroside
Empirical Formula (Hill Notation):
C
21
H
20
O
11
CAS Number:
522-12-3
Molecular Weight:
448.38
EC Number:
208-322-5
Products
Formula Weight
Manufacturer
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
analytical standard (1)
pharmaceutical primary standard (1)
primary reference standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
PHL89346
Quercitrin,
phyproof
®
Reference Substance
Expand
View Pricing
00740580
Quercitrin,
primary reference standard
Expand
View Pricing
Y0001931
Quercitrin,
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing