Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Q Sepharose
™
Fast Flow
Synonym(s):
Q Sepharose
™
Products
Brand
Sigma-Aldrich (1)
Manufacturer
Physical Form
Quality Segment
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
GE17-0510-01
Q Sepharose
™
Fast Flow,
Cytiva 17-0510-01, pack of 300 mL
Expand
View Pricing
Q1126
Q Sepharose
™
Fast Flow,
preswollen, 45-165 μm (wet), exclusion limit ~4,000,000 Da
Expand
View Pricing
GE17-0510-10
Q Sepharose
™
Fast Flow,
Cytiva 17-0510-10, pack of 25 mL
Expand
View Pricing