Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
serological pipettes, disposable, color-coded
Synonym(s):
Corning
®
7085
Products
Product Category
pipettes (8)
Manufacturer
Capacity (mL)
OD (mm)
Package Size
Sterility
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS708510
Pyrex
®
serological pipettes, disposable, color-coded,
volume 10.0 mL, accuracy: 0.06 mL
Expand
View Pricing
CLS70851
Pyrex
®
serological pipettes, disposable, color-coded,
volume 1.0 mL, accuracy: 0.02 mL, subdivision, 0.1 mL
Expand
View Pricing
CLS70855
Pyrex
®
serological pipettes, disposable, color-coded,
volume 5.0 mL, accuracy: 0.04 mL
Expand
View Pricing
CLS70851X
Pyrex
®
serological pipettes, disposable, color-coded,
volume 0.1 mL, accuracy: 0.005 mL
Expand
View Pricing
CLS70852
Pyrex
®
serological pipettes, disposable, color-coded,
volume 2.0 mL, accuracy: 0.02 mL, subdivision, 0.1 mL
Expand
View Pricing
CLS70852X
Pyrex
®
serological pipettes, disposable, color-coded,
volume 0.2 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS70852C
Pyrex
®
serological pipettes, disposable, color-coded,
volume 2.0 mL, accuracy: 0.02 mL, subdivision, 0.01 mL
Expand
View Pricing
CLS70851C
Pyrex
®
serological pipettes, disposable, color-coded,
volume 1.0 mL, accuracy: 0.02 mL, subdivision, 0.01 mL
Expand
View Pricing