Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
pipettes, Class A color-coded reusable
Products
Product Category
pipettes (11)
Color
Manufacturer
Capacity (mL)
Length (mm)
Glassware Class
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS710010
Pyrex
®
pipettes, Class A color-coded reusable,
volume 10 mL, accuracy: 0.02 mL
Expand
View Pricing
CLS71001
Pyrex
®
pipettes, Class A color-coded reusable,
volume 1 mL, accuracy: 0.006 mL
Expand
View Pricing
CLS710050
Pyrex
®
pipettes, Class A color-coded reusable,
volume 50 mL, accuracy: 0.05 mL
Expand
View Pricing
CLS71005
Pyrex
®
pipettes, Class A color-coded reusable,
volume 5 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS710025
Pyrex
®
pipettes, Class A color-coded reusable,
volume 25 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS71002
Pyrex
®
pipettes, Class A color-coded reusable,
volume 2 mL, accuracy: 0.006 mL
Expand
View Pricing
CLS710020
Pyrex
®
pipettes, Class A color-coded reusable,
volume 20 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS7100100
Pyrex
®
pipettes, Class A color-coded reusable,
volume 100 mL, accuracy: 0.08 mL
Expand
View Pricing
CLS71003
Pyrex
®
pipettes, Class A color-coded reusable,
volume 3 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS71004
Pyrex
®
pipettes, Class A color-coded reusable,
volume 4 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS710015
Pyrex
®
pipettes, Class A color-coded reusable,
volume 15 mL, accuracy: 0.03 mL
Expand
View Pricing