Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
petri dishes
Synonym(s):
Corning
®
3160
Products
Product Category
dishes (16)
Manufacturer
OD (mm)
Package Size
Sterility
Filter Products
Showing 1-16 of 16
Compare
Product Number
Description
Pricing
CLS316060
Pyrex
®
petri dishes,
complete, O.D. × H 60 mm × 15 mm
Expand
View Pricing
CLS3160102
Pyrex
®
petri dishes,
complete, O.D. × H 100 mm × 20 mm
Expand
View Pricing
CLS3160152
Pyrex
®
petri dishes,
complete, O.D. × H 150 mm × 20 mm
Expand
View Pricing
CLS3160102BO
Pyrex
®
petri dishes,
bottom only, O.D. × H 100 mm × 20 mm
Expand
View Pricing
CLS316060BO
Pyrex
®
petri dishes,
bottom only, O.D. × H 60 mm × 15 mm
Expand
View Pricing
CLS3160101BO
Pyrex
®
petri dishes,
bottom only, O.D. × H 100 mm × 17 mm
Expand
View Pricing
CLS3160152BO
Pyrex
®
petri dishes,
bottom only, O.D. × H 150 mm × 20 mm
Expand
View Pricing
CLS3160150BO
Pyrex
®
petri dishes,
bottom only, O.D. × H 150 mm × 15 mm
Expand
View Pricing
CLS3160100
Pyrex
®
petri dishes,
complete, O.D. × H 100 mm × 10 mm
Expand
View Pricing
CLS3160150
Pyrex
®
petri dishes,
complete, O.D. × H 150 mm × 15 mm
Expand
View Pricing
CLS3160101
Pyrex
®
petri dishes,
complete, O.D. × H 100 mm × 17 mm
Expand
View Pricing
CLS3160100CO
Pyrex
®
petri dishes,
cover only, O.D. × H 100 mm × 10 mm
Expand
View Pricing
CLS3160101CO
Pyrex
®
petri dishes,
cover only, O.D. × H 100 mm × 15 mm
Expand
View Pricing
CLS3160100BO
Pyrex
®
petri dishes,
bottom only, O.D. × H 100 mm × 10 mm
Expand
View Pricing
CLS316060CO
Pyrex
®
petri dishes,
cover only, O.D. × H 60 mm × 15 mm
Expand
View Pricing
CLS3160102CO
Pyrex
®
petri dishes,
cover only, O.D. × H 100 mm × 20 mm
Expand
View Pricing