Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
narrow-mouth graduated Erlenmeyer flask
Synonym(s):
Corning
®
4980
Products
Product Category
flasks (12)
Manufacturer
Capacity (mL)
Cap
Package Size
Sterility
Style
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
CLS4980125
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 125 mL
Expand
View Pricing
CLS4980250
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 250 mL
Expand
View Pricing
CLS49802L
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 2,000 mL
Expand
View Pricing
CLS4980500
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 500 mL
Expand
View Pricing
CLS498050
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 50 mL
Expand
View Pricing
CLS49801L
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 1,000 mL
Expand
View Pricing
CLS49806L
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 6,000 mL
Expand
View Pricing
CLS49804L
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 4,000 mL
Expand
View Pricing
CLS498025
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 25 mL
Expand
View Pricing
CLS49801XL
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 1,500 mL
Expand
View Pricing
CLS498010
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 10 mL, stopper joint: 00
Expand
View Pricing
CLS4980300
Pyrex
®
narrow-mouth graduated Erlenmeyer flask,
capacity 300 mL
Expand
View Pricing