Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PYREX
®
Griffin beakers
Synonym(s):
Corning
®
1000
Products
Product Category
beakers (15)
Brand
SAFC (3)
Manufacturer
Capacity (mL)
Package Size
Filter Products
Showing 1-15 of 15
Compare
Product Number
Description
Pricing
CLS1000PACK
PYREX
®
Griffin beakers,
low form, assorted pack (contains 1 beaker of 5 different sizes)
Expand
View Pricing
CLS1000600
PYREX
®
Griffin beakers,
Low form, capacity 600 mL
Expand
View Pricing
CLS10001L
PYREX
®
Griffin beakers,
Low form, capacity 1,000 mL
Expand
View Pricing
CLS1000250
PYREX
®
Griffin beakers,
Low form, capacity 250 mL
Expand
View Pricing
CLS10002L
PYREX
®
Griffin beakers,
Low form, capacity 2,000 mL
Expand
View Pricing
CLS100050
PYREX
®
Griffin beakers,
low form, capacity 50 mL
Expand
View Pricing
CLS1000100
PYREX
®
Griffin beakers,
low form, capacity 100 mL
Expand
View Pricing
CLS10004L
PYREX
®
Griffin beakers,
low form, capacity 4,000 mL
Expand
View Pricing
CLS1000150
PYREX
®
Griffin beakers,
low form, capacity 150 mL
Expand
View Pricing
CLS10003L
PYREX
®
Griffin beakers,
low form, capacity 3,000 mL
Expand
View Pricing
CLS1000400
PYREX
®
Griffin beakers,
Low form, capacity 400 mL
Expand
View Pricing
CLS100010
PYREX
®
Griffin beakers,
low form, capacity 10 mL
Expand
View Pricing
CLS100020
PYREX
®
Griffin beakers,
low form, capacity 20 mL
Expand
View Pricing
CLS100030
PYREX
®
Griffin beakers,
Low form, capacity 30 mL
Expand
View Pricing
CLS1000800
PYREX
®
Griffin beakers,
Low form, capacity 800 mL
Expand
View Pricing