Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations
Synonym(s):
Corning
®
3022
Products
Product Category
cylinders (8)
Manufacturer
Volume (ml)
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
CLS3022100
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 100 mL
Expand
View Pricing
CLS30221L
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 1,000 mL
Expand
View Pricing
CLS3022500
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 500 mL
Expand
View Pricing
CLS302225
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 25 mL
Expand
View Pricing
CLS30222L
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 2,000 mL
Expand
View Pricing
CLS302250
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 50 mL
Expand
View Pricing
CLS302210
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 10 mL
Expand
View Pricing
CLS3022250
Pyrex
®
graduated cylinder, single metric scale, white enamel graduations,
volume 250 mL
Expand
View Pricing