Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Pyrex
®
certified volumetric pipette, color-coded, Class A
Synonym(s):
Corning
®
7101
Products
Product Category
pipettes (11)
Color
Manufacturer
Capacity (mL)
Length (mm)
Glassware Class
Color
Package Size
Filter Products
Showing 1-11 of 11
Compare
Product Number
Description
Pricing
CLS710150
Pyrex
®
certified volumetric pipette, color-coded, Class A,
volume 50.0 mL, accuracy: 0.05 mL
Expand
View Pricing
CLS710115
Pyrex
®
certified volumetric pipette, color-coded, Class A,
green, volume 15.0 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS71011
Pyrex
®
certified volumetric pipette, color-coded, Class A,
blue, volume 1.0 mL, accuracy: 0.006 mL
Expand
View Pricing
CLS710110
Pyrex
®
certified volumetric pipette, color-coded, Class A,
red, volume 10.0 mL, accuracy: 0.02 mL
Expand
View Pricing
CLS71015
Pyrex
®
certified volumetric pipette, color-coded, Class A,
white, volume 5.0 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS7101100
Pyrex
®
certified volumetric pipette, color-coded, Class A,
volume 100.0 mL, accuracy: 0.08 mL
Expand
View Pricing
CLS71013
Pyrex
®
certified volumetric pipette, color-coded, Class A,
black, volume 3.0 mL, accuracy: 0.01 mL
Expand
View Pricing
CLS710120
Pyrex
®
certified volumetric pipette, color-coded, Class A,
yellow, volume 20.0 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS71012
Pyrex
®
certified volumetric pipette, color-coded, Class A,
orange, volume 2.0 mL, accuracy: 0.006 mL
Expand
View Pricing
CLS710125
Pyrex
®
certified volumetric pipette, color-coded, Class A,
blue, volume 25.0 mL, accuracy: 0.03 mL
Expand
View Pricing
CLS71014
Pyrex
®
certified volumetric pipette, color-coded, Class A,
red, volume 4.0 mL, accuracy: 0.01 mL
Expand
View Pricing