Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns
Products
Product Category
columns (9)
Manufacturer
pH Value
Product Line
Separation Technique
ID (mm)
Length (cm)
Package Size
Particle Size (µm)
Technique
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
Z777170
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 25 cm × 4.6 mm, stainless steel
Expand
View Pricing
Z777167
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 25 cm × 2.1 mm, stainless steel
Expand
View Pricing
Z777171
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 5 cm × 2.1 mm, PEEK
Expand
View Pricing
Z777168
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 1 cm × 4 mm, stainless steel
Expand
View Pricing
Z777173
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 5 cm × 4.6 mm, PEEK
Expand
View Pricing
Z777169
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 5 cm × 4.6 mm, stainless steel
Expand
View Pricing
Z777166
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 5 cm × 2.1 mm, stainless steel
Expand
View Pricing
Z777174
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 25 cm × 4.6 mm, PEEK
Expand
View Pricing
Z777172
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 25 cm × 2.1 mm, PEEK
Expand
View Pricing
Z777165
Proteomix
®
WCX-NP10 (10 µm) HPLC Columns,
L × I.D. 1 cm × 2 mm, stainless steel
Expand
View Pricing