Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Propyl 4-hydroxybenzoate
Synonym(s):
Propyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid propyl ester, Propyl paraben
Linear Formula:
HOC
6
H
4
CO
2
CH
2
CH
2
CH
3
CAS Number:
94-13-3
Molecular Weight:
180.20
EC Number:
202-307-7
Products
Product Category
building blocks (1)
preservatives (1)
Brand
SAFC (1)
Sigma-Aldrich (3)
Feature
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
1.07427
Propyl 4-hydroxybenzoate,
EMPROVE
®
EXPERT, Ph. Eur., BP, ChP, JP, NF
Expand
View Pricing
P53357
Propyl 4-hydroxybenzoate,
≥99%
Expand
View Pricing
P5835
Propyl 4-hydroxybenzoate,
BioXtra
Expand
View Pricing
54789
Propyl 4-hydroxybenzoate,
tested according to Ph. Eur.
Expand
View Pricing