Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
PrepSafe
™
microcentrifuge tube mini floating rack
Products
Product Category
racks (8)
Manufacturer
Diameter (mm)
OD (mm)
Color
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
Z756385
PrepSafe
™
microcentrifuge tube mini floating rack,
clear
Expand
View Pricing
Z756431
PrepSafe
™
microcentrifuge tube mini floating rack,
with vortex attachment, turquoise
Expand
View Pricing
Z756407
PrepSafe
™
microcentrifuge tube mini floating rack,
pink (fuchsia)
Expand
View Pricing
Z756393
PrepSafe
™
microcentrifuge tube mini floating rack,
turquoise
Expand
View Pricing
Z756415
PrepSafe
™
microcentrifuge tube mini floating rack,
yellow (neon)
Expand
View Pricing
Z756423
PrepSafe
™
microcentrifuge tube mini floating rack,
clear
Expand
View Pricing
Z756458
PrepSafe
™
microcentrifuge tube mini floating rack,
pink (fuchsia)
Expand
View Pricing
Z756466
PrepSafe
™
microcentrifuge tube mini floating rack,
yellow (neon)
Expand
View Pricing