Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium oxalate monohydrate
Synonym(s):
Ethanedioic acid, Oxalic acid potassium salt
Linear Formula:
(COOK)
2
· H
2
O
CAS Number:
6487-48-5
Molecular Weight:
184.23
EC Number:
209-506-8
Products
Product Category
building blocks (3)
salts (1)
Brand
Sigma-Aldrich (3)
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
ACS reagent (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
223425
Potassium oxalate monohydrate,
ACS reagent, 99%
Expand
View Pricing
P0963
Potassium oxalate monohydrate,
BioXtra, ≥98.5%
Expand
View Pricing
24-5070
Potassium oxalate monohydrate,
JIS special grade, ≥99.0%
Expand
View Pricing