Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Potassium borohydride
Synonym(s):
Potassium borohydride, Potassium tetrahydroborate
Linear Formula:
KBH
4
CAS Number:
13762-51-1
Molecular Weight:
53.94
EC Number:
237-360-5
Products
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Special Grade
Purum (1)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
8.20747
Potassium borohydride,
for synthesis
Expand
View Pricing
455571
Potassium borohydride,
≥97%
Expand
View Pricing
438472
Potassium borohydride,
99.9% trace metals basis
Expand
View Pricing
60080
Potassium borohydride,
purum, ≥97.0% (RT)
Expand
View Pricing