Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(vinyl acetate)
Synonym(s):
Poly(vinyl acetate)
Linear Formula:
[CH
2
CH(O
2
CCH3)]
n
CAS Number:
9003-20-7
Products
Brand
Sigma-Aldrich (2)
Manufacturer
Physical Form
Format
Quality Segment
Application
Polymer Composition
Polymer Type
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (2)
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
189480
Poly(vinyl acetate),
average M
w
~100,000 by GPC, beads
Expand
View Pricing
387932
Poly(vinyl acetate),
average M
w
~500,000 by GPC
Expand
View Pricing
1548032
Poly(vinyl acetate),
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
Y0001557
Poly(vinyl acetate),
European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing