
Poly(propylene glycol)-block-poly(ethylene glycol)-block-poly(propylene glycol)

Synonym(s):
PPG-PEG-PPG
CAS Number:
Products
Brand
Physical Form
Chemical Composition
Quality Segment
Polymer Composition
Polymer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing