Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(dimethylsiloxane), bis(3-aminopropyl) terminated
Synonym(s):
PDMS
Linear Formula:
H
2
N(CH
2
)
3
Si(CH
3
)
2
O[Si(CH
3
)
2
O]
n
Si(CH
3
)
2
(CH
2
)
3
NH
2
CAS Number:
106214-84-0
Products
Boiling Point (°C)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
481688
Poly(dimethylsiloxane), bis(3-aminopropyl) terminated,
average M
n
~2,500
Expand
View Pricing
481696
Poly(dimethylsiloxane), bis(3-aminopropyl) terminated,
average M
n
~27,000
Expand
View Pricing