Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Poly(2-hydroxyethyl methacrylate)
Synonym(s):
Poly(2-HEMA), Poly-HEMA
Linear Formula:
(C
6
H
10
O
3
)
n
CAS Number:
25249-16-5
Products
Biological Source
Physical Form
Product Line
Technique
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
P3932
Poly(2-hydroxyethyl methacrylate),
BioReagent, powder, suitable for cell culture
Expand
View Pricing
192066
Poly(2-hydroxyethyl methacrylate),
average M
v
300,000, crystalline
Expand
View Pricing
529265
Poly(2-hydroxyethyl methacrylate),
average M
v
20,000
Expand
View Pricing
529257
Poly(2-hydroxyethyl methacrylate),
average M
v
1,000,000
Expand
View Pricing