Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum-tungsten alloy, Pt92W8
Products
Manufacturer
Physical Form
Diameter (mm)
Length (m)
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
GF36897395
Platinum-tungsten alloy, Pt92W8,
20m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF99176732
Platinum-tungsten alloy, Pt92W8,
5m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF02174217
Platinum-tungsten alloy, Pt92W8,
1m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation
Expand
View Pricing
GF18205107
Platinum-tungsten alloy, Pt92W8,
10m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF32280533
Platinum-tungsten alloy, Pt92W8,
50m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF55448365
Platinum-tungsten alloy, Pt92W8,
20m wire, 0.025mm diameter, as drawn
Expand
View Pricing
GF89020605
Platinum-tungsten alloy, Pt92W8,
2m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation
Expand
View Pricing
GF47238374
Platinum-tungsten alloy, Pt92W8,
10m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation
Expand
View Pricing
GF01100574
Platinum-tungsten alloy, Pt92W8,
20m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation
Expand
View Pricing
GF25345920
Platinum-tungsten alloy, Pt92W8,
5m insulated wire, 0.025mm conductor diameter, 0.007mm insulation thickness, polyimide insulation
Expand
View Pricing
GF97866084
Platinum-tungsten alloy, Pt92W8,
5m wire, 0.05mm diameter, as drawn
Expand
View Pricing
GF24497365
Platinum-tungsten alloy, Pt92W8,
10m wire, 0.05mm diameter, as drawn
Expand
View Pricing