Skip to Content
AI
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Platinum on carbon
Empirical Formula (Hill Notation):
Pt
Molecular Weight:
195.08
Products
Brand
Sigma-Aldrich (12)
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Reagent Type
Core
Filter Products
Showing 1-12 of 12
Compare
Product Number
Description
Pricing
205958
Platinum on carbon,
extent of labeling: 10 wt. % loading, matrix activated carbon support
Expand
View Pricing
205931
Platinum on carbon,
extent of labeling: 5 wt. % loading, matrix activated carbon support
Expand
View Pricing
330159
Platinum on carbon,
Evonik NOBLYST
®
P2056 5% Pt
Expand
View Pricing
907898
Platinum on carbon,
Evonik Noblyst
®
P8078 1% Pt
Expand
View Pricing
237558
Platinum on carbon,
extent of labeling: 3 wt. % loading, matrix activated carbon support
Expand
View Pricing
908037
Platinum on carbon,
Evonik Noblyst
®
P2061 5% Pt
Expand
View Pricing
908150
Platinum on carbon,
Evonik Noblyst
®
P2060 5% Pt
Expand
View Pricing
907952
Platinum on carbon,
Evonik Noblyst
®
P8071 2% Pt
Expand
View Pricing
908088
Platinum on carbon,
Evonik Noblyst
®
P2065 3% Pt
Expand
View Pricing
907812
Platinum on carbon,
Evonik Noblyst
®
P2086 10% Pt
Expand
View Pricing
908053
Platinum on carbon,
Evonik Noblyst
®
P2058 5% Pt
Expand
View Pricing
908169
Platinum on carbon,
Evonik Noblyst
®
P2057 3% Pt
Expand
View Pricing